missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Sodium Butyrate

Product Code. 17180042
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17180042 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17180042 Supplier STEMCELL Technologies Supplier No. 72242
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Epigenetic modifier; Inhibits histone deacetylase

  • Sodium Butyrate is the sodium salt of butyric acid, a short chain fatty acid that inhibits histone deacetylases (HDACs), leading to hyperacetylation of histones
  • This causes changes in chromatin structure and gene expression, resulting in many biological effects
  • (Boffa et al.; Kruh)
  • Promotes reprogramming of human somatic cells to induced pluripotent stem (iPS) cells using only a single factor, OCT4 (Zhu et al.)
  • Supports self-renewal of mouse and human embryonic stem (ES) cells, in the absence of exogenously added growth factors (Ware et al.)
  • Promotes differentiation to hepatocytes from mouse and human ES cells (Hay et al.; Zhou et al.)
  • Promotes differentiation to definitive endoderm and islet-like cells from human ES cells (Jiang et al.)
  • Enhances osteogenic and suppresses adipogenic differentiation from human mesenchymal cells (Chen et al.; Lee et al.)

Specifications

CAS 156-54-7
Synonym Butanoic acid; Butyric acid, sodium salt
Quantity 500 mg
Molecular Formula C4H7O2 · Na
Purity ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.