missing translation for 'onlineSavingsMsg'
Learn More

Sodium bromoacetate, 98%

Product Code. 11378096
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378096 25 g 25g
11368096 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378096 Supplier Thermo Scientific Alfa Aesar Supplier No. L14983.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1068-52-6 | C2H2BrNaO2 | 160.93 g/mol

It is an important raw material and intermediate used in organic synthesis, agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1068-52-6
Molecular Formula C2H2BrNaO2
Molecular Weight (g/mol) 160.93
MDL Number MFCD01311832
InChI Key SESSOVUNEZQNBV-UHFFFAOYSA-M
Synonym sodium bromoacetate, sodium 2-bromoacetate, sodium monobromoacetate, unii-8j155ck8pv, acetic acid, bromo-, sodium salt, bromoacetic acid sodium salt, acmc-1bowb, sodium bromoacetate, ?brch2coona, acetic acid,bromo-,sodium salt 8ci,9ci, acetic acid, bromo-,sodium salt 8ci,9ci
PubChem CID 23675628
IUPAC Name sodium;2-bromoacetate
SMILES C(C(=O)[O-])Br.[Na+]

Specifications

Melting Point >300°C
Odor Odorless
Quantity 25 g
UN Number UN2811
Solubility Information Soluble in water.
Formula Weight 160.93
Percent Purity 98%
Chemical Name or Material Sodium bromoacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.