missing translation for 'onlineSavingsMsg'
Learn More

Sodium bromide, ultra dry, 99.999% (metals basis)

Product Code. 11388208
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388208 25 g 25g
11378208 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388208 Supplier Thermo Scientific Alfa Aesar Supplier No. 039426.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7647-15-6 | BrNa | 102.89 g/mol

Sodium bromide is used in the preparation of bromides and as a source of bromide ion. It also finds its applications in drilling fluid additives, and weighting agent. It is employed in , Oil gas production, hydraulic fracturing, and fracking. It is also used in photography, to make bromides and organic chemicals, as a therapeutic sedative and hypnotic, and formerly as a veterinary medicine. It also finds its applications to control algae, bacteria, and fungi in water systems (pasteurizing, canning, pulp & paper manufacturing, and ornamental ponds); and used to repel moths from clothing and fleas from pets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
PubChem CID 253881
ChEBI CHEBI:63004
IUPAC Name sodium bromide
SMILES [Na+].[Br-]

Specifications

Boiling Point 1390°C
Melting Point 755°C
Physical Form Spheres
Quantity 25 g
Merck Index 14,8594
Solubility Information Soluble in water at 20°C 900g/L,alcohol (62.5mg/ml),methanol (166.67mg/ml),and 95% ethanol (70mg/ml).
Formula Weight 102.89
Percent Purity 99.999%
Packaging Ampule
Refractive Index 1.6412
Sensitivity Hygroscopic
Density 3.203 g/mL
Assay (metals basis)
Chemical Name or Material Sodium bromide, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.