missing translation for 'onlineSavingsMsg'
Learn More

Sodium bromide, ACS reagent

Product Code. 12035350 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
12035350 500 g Glass bottle 500g
12025350 100 g Glass bottle 100g
12015350 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12035350 Supplier Thermo Scientific Acros Supplier No. 212675000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7647-15-6 | BrNa | 102.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
PubChem CID 253881
ChEBI CHEBI:63004
SMILES [Na+].[Br-]

Specifications

Boiling Point 1390.0°C
Melting Point 755.0°C
Quantity 500 g
Linear Formula NaBr
Fieser 12,445; 13,46
Merck Index 15,8729
Solubility Information Solubility in water: 905g/L (20°C). Other solubilities: 1160g/L water (50°C), 1210g/L water (100°C), 1g/6 mL methanol, 7g/100 mL 95% ethanol
IUPAC Name sodium bromide
Formula Weight 102.89
Grade ACS Reagent
Packaging Glass bottle
Chemical Name or Material Sodium bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.