missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium bromide, 99+%, for biochemistry

Product Code. 12123761 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
12123761 500 g Plastic bottle 500g
12113761 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12123761 Supplier Thermo Scientific Chemicals Supplier No. 446805000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7647-15-6 | BrNa | 102.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
PubChem CID 253881
ChEBI CHEBI:63004
SMILES [Na+].[Br-]

Specifications

Boiling Point 1390.0°C
Melting Point 755.0°C
Quantity 500 g
Assay Percent Range 99% min. (Argentometry)
Linear Formula NaBr
Merck Index 15,8729
Solubility Information Solubility in water: 905g/L (20°C). Other solubilities: 1160g/L water (50°C), 1210g/L water (100°C), 1g/6 mL methanol, 7g/100 mL 95% ethanol
IUPAC Name sodium bromide
Formula Weight 102.89
Percent Purity ≥99%
Grade Biochemical
Packaging Plastic bottle
Chemical Name or Material Sodium bromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium bromide
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H336-May cause drowsiness or dizziness.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Sodium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.