missing translation for 'onlineSavingsMsg'
Learn More

Sodium bromide, Puratronic™, 99.9955% (metals basis), Thermo Scientific Chemicals

Product Code. 11346419
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Sodium bromide is widely used for the preparation of other bromides in organic synthesis and other areas. Also used to prepare dense fluids used in oil wells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
PubChem CID 253881
ChEBI CHEBI:63004
SMILES [Na+].[Br-]

Specifications

Boiling Point 1390°C
Melting Point 755°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.9955% (metals basis)
Solubility Information Soluble in water. Also soluble in alcohol,methanol and ethanol.
IUPAC Name sodium bromide
Formula Weight 102.89
Refractive Index 1.6412
Sensitivity Hygroscopic
Appearance Powder/flake/crystalline/beads
Density 3.203 g/cm3 at 20°C
Chemical Name or Material Sodium bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'