missing translation for 'onlineSavingsMsg'
Learn More

Sodium bromide, 99.99% (metals basis)

Product Code. 11334989
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334989 25 g 25g
11324989 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334989 Supplier Thermo Scientific Alfa Aesar Supplier No. 087604.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7647-15-6 | BrNa | 102.89 g/mol

Sodium bromide is used in pharmaceutical applications, used in photographic processing and chemical intermediate for production of chemicals and bromides. It is widely used as an anticonvulsant. Also used for the preparation of drilling fluids, completion fluids and drilling muds formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
PubChem CID 253881
ChEBI CHEBI:63004
SMILES [Na+].[Br-]

Specifications

Boiling Point 1390°C
Melting Point 755°C
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in water and alcohol.
IUPAC Name sodium;bromide
Formula Weight 102.89
Refractive Index 1.6412
Sensitivity Hygroscopic
Density 3.203
Chemical Name or Material Sodium bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.