missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bromate, Solstice

Product Code. 15610580 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 g
1 kg
25 kg
Unit Size:
1kg
250g
25kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15610580 25 kg 25kg
15600580 250 g 250g
15690570 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15610580 Supplier Honeywell-Fluka Supplier No. 0215125KG
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-38-0
Molecular Formula BrNaO3
Molecular Weight (g/mol) 150.891
MDL Number MFCD00003476
InChI Key XUXNAKZDHHEHPC-UHFFFAOYSA-M
Synonym sodium bromate, bromic acid, sodium salt, dyetone, bromate de sodium, sodium bromate dot, neutralizer k-126, neutralizer k-140, neutralizer k-938, sodium bromate nabro3, bromate de sodium french
PubChem CID 23668195
ChEBI CHEBI:75229
IUPAC Name sodium;bromate
SMILES [O-]Br(=O)=O.[Na+]

Specifications

Physical Form Crystalline Powder
Quantity 25 kg
Linear Formula NaBrO3
UN Number 1494
Formula Weight 150.89g/mol
Percent Purity ≥99%
Density 3.339g/mL at 25°C
Chemical Name or Material Sodium Bromate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.