missing translation for 'onlineSavingsMsg'
Learn More

Sodium Borohydride, TRC

Artikelnummer. 30410162
Ansicht ändern
Click to view available options
Quantity:
250 g
Packungsgröße:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packungsgröße
30410162 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Abgabe sachkundepflichtiger Stoffe und Gemische nur bei Erfüllung der Vorrausetzungen nach Chemikalienverbotsverordnung (ChemVerbotsV). Keine Abgabe an Privatpersonen.
Product Code. 30410162 Supplier Toronto Research Chemicals Supplier No. TRCS614900250G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Borohydride
CAS 16940-66-2
Quantity 250 g
Molecular Formula BH4Na
Synonym Tetrahydroborate(1-) Sodium, Borol, Colorstrip 2000, Hidkitex DF, Hydrifin, Sodium Borohydrate, Sodium Borohydride, Sodium Borohydride, NaBH4, Sodium Hydroborate, Sodium Tetrahydridoborate, Sodium tetrahydroborate, Sodium Tetrahydroborate(1-), VenMet
InChI Formula InChI=1S/BH4.Na/h1H4;/q-1;+1
SMILES [BH4-].[Na+]
IUPAC Name sodium;boranuide
Molecular Weight (g/mol) 37.83
Formula Weight 38.0304
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.