missing translation for 'onlineSavingsMsg'
Learn More

Sodium Borohydride, Technical, SLR, Fisher Chemical™

Product Code. 10599010 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10599010 25 g 25g
10409190 10 g 10g
10703621 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
Product Code. 10599010 Supplier Fisher Chemical Supplier No. S/2560/46
  • ONLINE EXCLUSIVE PRICE
    £119.00
    £78.90 / 25g
    Save £40.10
    33% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Chemical Identifiers

CAS 16940-66-2
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003518
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]

Specifications

Melting Point 360°C
Color White
pH 11
Physical Form Powder/Solid
Quantity 25 g
Packaging Plastic powder jar
Formula Weight 37.83g/mol
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium borohydride
Signal Word
  • Danger
Hazard Category
  • Substance which, in contact with water, emits flammable gases Category 1
  • Skin corrosion/irritation Category 1 C
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 3
  • Reproductive toxicity Category 1B
Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H360FD-May damage fertility. May damage the unborn child.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P223-Do not allow contact with water.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.