missing translation for 'onlineSavingsMsg'
Learn More

Sodium borohydride, stable aq. soln., 4.4M in 14M NaOH, Thermo Scientific Chemicals

Catalog Number p-7080365
Click to view available options
Quantity:
100 mL
500 mL
2 L
This item is not returnable. View return policy
16940-66-2
BH4Na
37.83
MFCD00003518
YOQDYZUWIQVZSF-UHFFFAOYSA-N
Sodium tetrahydridoborate
sodium boranuide
[BH4-].[Na+]
This item is not returnable. View return policy
16940-66-2
BH4Na
37.83
MFCD00003518
YOQDYZUWIQVZSF-UHFFFAOYSA-N
Sodium tetrahydridoborate
sodium boranuide
[BH4-].[Na+]

Sodium borohydride is used as a reagent in the reduction of amino acids and their derivatives. Also used as a catalyst in ammonia borane dehydrogenation. It acts as a reducing agent in the synthesis of gold nanoparticles. It is also used for wood pulp and as a blowing agent for plastics. It reduces aldehyde and keto groups to give the corresponding alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Physical Form Liquid
Quantity 100 mL
Linear Formula NaBH4
UN Number UN3320
Solubility Information Miscible with water,liquid ammonia,amines and pyridine.
Formula Weight 37.83
Concentration or Composition (by Analyte or Components) 4.4M in 14M NaOH
Sensitivity Air Sensitive
Density 1.4 g/mL
Chemical Name or Material Sodium borohydride, Stable aq. soln.
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'