missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium borohydride, stable aq. soln., 4.4M in 14M NaOH, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
2 L
Description
Sodium borohydride is used as a reagent in the reduction of amino acids and their derivatives. Also used as a catalyst in ammonia borane dehydrogenation. It acts as a reducing agent in the synthesis of gold nanoparticles. It is also used for wood pulp and as a blowing agent for plastics. It reduces aldehyde and keto groups to give the corresponding alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 100 mL |
Linear Formula | NaBH4 |
UN Number | UN3320 |
Solubility Information | Miscible with water,liquid ammonia,amines and pyridine. |
Formula Weight | 37.83 |
Concentration or Composition (by Analyte or Components) | 4.4M in 14M NaOH |
Sensitivity | Air Sensitive |
Density | 1.4 g/mL |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'