Learn More
Sodium borohydride, 12% solution in 40% aq. sodium hydroxide solution, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 389930025
Code : 29
Additional Details : CAS Number : 16940-66-2 Weight : 3.50000kg Transport : UN number : 3320 Chem class : 8 Pack group : II
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
130.0°C to 135.0°C | |
12% solution in 40% aq. sodium hydroxide solution | |
Colorless | |
2.5 L | |
NaBH4 | |
15, 8727 | |
Solubility in water: completely soluble | |
[BH4-].[Na+] | |
37.83 | |
11 to 13% | |
1.4 |
Sodium borohydride | |
16940-66-2,1310-73-2,67-56-1,7732-18-5 | |
Liquid | |
BH4Na | |
MFCD00003518 | |
Sodium tetrahydroborate, SBH | |
YOQDYZUWIQVZSF-UHFFFAOYSA-N | |
sodium boranuide | |
37.82 | |
Plastic bottle | |
1.4000g/mL |
RUO – Research Use Only