missing translation for 'onlineSavingsMsg'
Learn More

Sodium borohydride, 12% solution in 40% aq. sodium hydroxide solution

Product Code. 10413205 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
1 L
2.5 L
Packaging:
Plastic bottle
Unit Size:
1 L
100mL
2.5 L
250mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10413205 1 L Plastic bottle 1 L
10310424 100 mL Plastic bottle 100mL
10248893 250 mL Plastic bottle 250mL
11360836 2.5 L Plastic bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10413205 Supplier Thermo Scientific Supplier No. 389930010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16940-66-2 | BH4Na | 37.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 130.0°C to 135.0°C
Chemical Name or Material Sodium borohydride
Name Note 12% solution in 40% aq. sodium hydroxide solution
CAS 16940-66-2, 1310-73-2, 67-56-1, 7732-18-5
Color Colorless
Physical Form Liquid
Quantity 1 L
Molecular Formula BH4Na
Linear Formula NaBH4
MDL Number MFCD00003518
Merck Index 15, 8727
Synonym Sodium tetrahydroborate, SBH
Solubility Information Solubility in water: completely soluble.
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
SMILES [BH4-].[Na+]
IUPAC Name sodium boranuide
Molecular Weight (g/mol) 37.83
Formula Weight 37.82
Percent Purity 11 to 13%
Packaging Plastic bottle
Specific Gravity 1.4
Density 1.4000g/mL
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.