missing translation for 'onlineSavingsMsg'
Learn More

Sodium borohydride, 12% solution in 40% aq. sodium hydroxide solution, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 389931000

 View more versions of this product

Product Code. 10310424

  • £33.20 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY
Specifications

Specifications

130.0°C to 135.0°C
12% solution in 40% aq. sodium hydroxide solution
Colorless
100 mL
NaBH4
15, 8727
Solubility in water: completely soluble
[BH4-].[Na+]
37.83
11 to 13%
1.4
Sodium borohydride
16940-66-2, 1310-73-2, 67-56-1, 7732-18-5
Liquid
BH4Na
MFCD00003518
Sodium tetrahydroborate, SBH
YOQDYZUWIQVZSF-UHFFFAOYSA-N
sodium boranuide
37.82
Plastic bottle
1.4000g/mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only