missing translation for 'onlineSavingsMsg'
Learn More

Sodium borohydride, 20% aq. NaOH solution, ACROS Organics™

Product Code. p-4839198 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Unit Size:
100mL
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 13455122

Brand: Thermo Scientific Acros 448521000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 16940-66-2, 1310-73-2
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003518
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]

Specifications

Boiling Point 120.0°C to 125.0°C
Quantity 100 mL
Solubility Information Solubility in water: completely soluble.
Formula Weight 37.83
Packaging Plastic bottle
Specific Gravity 1.2
Density 1.2000g/mL
Chemical Name or Material Sodium borohydride, 20% aq. NaOH solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.