missing translation for 'onlineSavingsMsg'
Learn More

Sodium borohydride, 20% aq. NaOH solution, ACROS Organics™

Product Code. 13455122 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13455122 100 mL 100mL
13465122 500 mL 500mL
13445122 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13455122 Supplier Thermo Scientific Acros Supplier No. 448521000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 16940-66-2 , 1310-73-2
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003518
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]

Specifications

Boiling Point 120.0°C to 125.0°C
Quantity 100 mL
Solubility Information Solubility in water: completely soluble.
Formula Weight 37.83
Packaging Plastic bottle
Specific Gravity 1.2
Density 1.2000g/mL
Chemical Name or Material Sodium borohydride, 20% aq. NaOH solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.