missing translation for 'onlineSavingsMsg'
Learn More

Sodium Borohydride, 0.98, BAKER, J.T. Baker™

Product Code. 13275503 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
13275503 100 g Poly Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13275503 Supplier Avantor J.T.Baker Supplier No. V02305

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A grade of chemicals for laboratory use

    For Laboratory,Research,or Manufacturing Use

Specifications

CAS 16940-66-2
Quantity 100 g
Molecular Formula BH4Na
MDL Number MFCD00003518
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
SMILES [BH4-].[Na+]
IUPAC Name sodium boranuide
Molecular Weight (g/mol) 37.83
Formula Weight 37.83
Percent Purity 98%
Packaging Poly Bottle
Specific Gravity 1L = 1.07kg
Chemical Name or Material Sodium Borohydride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.