missing translation for 'onlineSavingsMsg'
Learn More

Sodium borodeuteride, 99% (Isotopic)

Product Code. 11347798
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347798 1 g 1g
11337798 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11347798 Supplier Thermo Scientific Alfa Aesar Supplier No. 035102.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 15681-89-7 | BH4Na | 37.83 g/mol

Sodium borodeuteride is used to prepare alfa, alfa-dideutero-beta-amino acids. It is also used to prepare amine-stabilized, deuterated borane-tetrhydrofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15681-89-7
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003519
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
Synonym sodium borodeuteride, sodium tetradeuterioboron, sodium borohydride-d4, sodium 2h4 tetrahydroborate 1-, sulfatase, borate 1-, tetrahydro-d4-, sodium, nabd4, sodium 2h4 boranuide, deuterated sodium borohydride
PubChem CID 23673181
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]

Specifications

Physical Form Powder
Color White
Sensitivity Moisture
Quantity 1 g
pH 11
Melting Point ∼400°C (decomposition)
Linear Formula NaBD4
UN Number UN1426
Solubility Information Soluble in liquid ammonia,amines and pyridine.
Formula Weight 41.88
Percent Purity 99%
Chemical Name or Material Sodium borodeuteride, Isotopic
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium borodeuteride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Substance which, in contact with water, emits flammable gases Category 2
Hazard Statement
  • H261-In contact with water releases flammable gases.
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P223-Do not allow contact with water.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Sodium borodeuteride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.