missing translation for 'onlineSavingsMsg'
Learn More

Sodium Borate, Decahydrate, MP Biomedicals™

Product Code. 11414642 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414642 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11414642 Supplier MP Biomedicals Supplier No. 02191436.5
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in biochemical and chemical laboratories to make buffers. Sodium Borate, Decahydrate, MP Biomedicals is a mineral and salt of boric acid, often referred to borax. It is also used as a macronutrient fertilizer and to clean skills in taxidermy.

The term borax is used for a number of closely related minerals or chemical compounds that differ in their crystal water content, but usually refers to the decahydrate. Sodium borate is used for many applications:

  • To make buffers, e.g. for gel electrophoresis of DNA, such as TBE (Tris/Borate/EDTA) or SB (Sodium borate) buffer, or as BBS (borate buffered saline) in coating procedures
  • As a micronutrient fertilizer to correct boron-deficient soils
  • To clean the brain cavity of a skull for mounting

Specifications

CAS 1303-96-4
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 99.5–105.0%
Molecular Formula B4Na2O7.10H2O
Solubility Information 1g dissolves in 16M water, 0.6mL boiling water, ∼/mL glyceral; insoluble in alcohol.
Molecular Weight (g/mol) 381.37
Formula Weight 381.37g/mol
Grade Not Available
Chemical Name or Material Sodium Borate Decahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.