missing translation for 'onlineSavingsMsg'
Learn More

Sodium Borate Decahydrate, 99.5-102.0%, (Crystalline/ACS Reagent Grade), MP Biomedicals

Product Code. 11434642 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 kg
Unit Size:
5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434642 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11434642 Supplier MP Biomedicals Supplier No. 0219143605
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1303-96-4
Physical Form Crystalline
Quantity 5 kg
Assay Percent Range 99.5-102.0%
Molecular Formula Na2B4O7 · 10H2O
Formula Weight 381.37
Grade ACS Reagent
Chemical Name or Material Sodium Borate Decahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.