missing translation for 'onlineSavingsMsg'
Learn More

Sodium Borate, 10-Hydrate, N.F., J.T. Baker™

Catalog Number p-5390881
195.70 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
1303-96-4
Na2B4O7·10H2O
This item is not returnable. View return policy
1303-96-4
Na2B4O7·10H2O
Quantity 500 g
Formula Weight 381.37
Grade NF
Packaging Poly Bottle
Specific Gravity 1L = 1.73kg
Chemical Name or Material Sodium Borate, 10-Hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'