missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bitartrate Monohydrate, TRC

Product Code. 30411865
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30411865 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411865 Supplier Toronto Research Chemicals Supplier No. TRCS083928100G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Bitartrate Monohydrate
CAS 6131-98-2
Molecular Formula C4 H5 O6 . Na . H2 O
Quantity 100 g
Synonym Sodium Hydrogen Tartrate Monohydrate, (2R,3R)-2,3-Dihydroxybutanedioic Acid Sodium Salt Monohydrate, Sodium Tartaric Acid Monohydrate
InChI Formula InChI=1S/C4H6O6.Na.H2O/c5-1(3(7)8)2(6)4(9)10;;/h1-2,5-6H,(H,7,8)(H,9,10);;1H2/q;+1;/p-1/t1-,2-;;/m1../s1
SMILES O.[Na+].O[C@H]([C@@H](O)C(=O)[O-])C(=O)O
IUPAC Name sodium;(2R,3R)-2,3,4-trihydroxy-4-oxobutanoate;hydrate
Molecular Weight (g/mol) 190.084
Formula Weight 190.009
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.