missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bisulfite (mixture of NaHSO3 and Na2S2O5), TRC

Product Code. 30010486
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010486 25 g 25g
30005389 5 g 5g
30011069 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010486 Supplier Toronto Research Chemicals Supplier No. TRCS61440025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-90-5
Formula Weight 103.9544
InChI Formula InChI=1S/Na.H2O3S/c;1-4(2)3/h;(H2,1,2,3)/q+1;/p-1
IUPAC Name sodium;hydrogen sulfite
Molecular Formula NaHSO3
Molecular Weight (g/mol) 104.06
SMILES [Na+].OS(=O)[O-]
Synonym Sulfurous Acid Monosodium Salt, E 222, FR-62, Hydrogen Sodium Sulfite, Hydrogen Sulfite Sodium, Monosodium Sulfite, Sodium Acid Sulfite, Sodium Bisulfite, Sodium Bisulfite (NaHSO3), Sodium Bisulphite, Sodium Hydrogen Sulfite, Sodium Sulfite (NaHSO3);
Chemical Name or Material Sodium Bisulfite (mixture of NaHSO3 and Na2S2O5)

Specifications

Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.