missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bisulfite ACS AR Granular, Macron Fine Chemicals™

Product Code. p-3178131
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly
Unit Size:
2.5kg
This item is not returnable. View return policy

Product Code. 15538754

Brand: Avantor Macron 744806

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7631-90-5
Molecular Formula NaO3S
Molecular Weight (g/mol) 103.05
MDL Number MFCD00003530
InChI Key DWAQJAXMDSEUJJ-UHFFFAOYSA-L
Synonym sodium bisulfite, sodium hydrogen sulfite, sodium bisulphite, sodium hydrogensulfite, sodium sulfhydrate, monosodium sulfite, uantax sbs, sulfurous acid, monosodium salt, caswell no. 750, hydrogen sodium sulfite
PubChem CID 23665763
ChEBI CHEBI:26709
IUPAC Name sodium sulfite
SMILES [Na+].[O-]S([O-])=O

Specifications

Physical Form Granules
Quantity 2.5 kg
Formula Weight 104.06
Grade ACS
Packaging Poly
Chemical Name or Material Sodium Bisulfite
compliance-icons compliance-icons
Product Identifier
  • Sodium hydrogen sulphite
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Sodium hydrogen sulphite
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.