missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bisulfate Monohydrate, 99%, ReagentPlus™, Honeywell™

Product Code. 15612320 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly Bottle
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15612320 100 g Poly Bottle 100g
15632320 500 g Poly Bottle 500g
15622320 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15612320 Supplier Honeywell Research Chemicals Supplier No. 233714100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-88-5
Molecular Formula H3NaO5S
Molecular Weight (g/mol) 138.07
MDL Number MFCD00149210
InChI Key JXHZRQHZVYDRGX-UHFFFAOYSA-M
Synonym sodium bisulfate monohydrate, sodium hydrogen sulfate monohydrate, unii-3kls9y77yj, sulfuric acid, monosodium salt, hydrate, 3kls9y77yj, sodium hydrogensulfate hydrate, sulfuric acid, monosodium salt, monohydrate, sodium hydrate hydrogen sulfate, sodium hydrogen sulfate-1-hydrate, acmc-20ajox
PubChem CID 23673662
IUPAC Name sodium hydrate hydrogen sulfate
SMILES O.[Na+].OS([O-])(=O)=O

Specifications

Melting Point 58°C
Color White
Quantity 100 g
Linear Formula NaHSO4 · H2O
UN Number NONH for all modes of transport
Formula Weight 138.08g/mol
Percent Purity 99%
Grade ReagentPlus
Packaging Poly Bottle
Appearance Moist crystals
Density 2.12g/cm3
Chemical Name or Material Sodium Bisulfate Monohydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.