missing translation for 'onlineSavingsMsg'
Learn More

Sodium bismuth oxide, 80%

Product Code. p-7021656
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11428006

Brand: Thermo Scientific Alfa Aesar A14073.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12232-99-4 | BiNaO3 | 279.967 g/mol

Sodium bismuthate oxide is used as an oxidant in the preparation of hexavalent Americium (Am) during the separation of Am from the lanthanides., hydrothermal synthesis of bismuth oxychloride (BiOCl) micrometer-sized single crystals. It is also used as a precursor in the synthesis of bismuth nanoforms under microwave (MW) irradiation. It used to converts almost any manganese compound to permanganate, which is easily assayed spectrophotometrically.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium bismuthate oxide is used as an oxidant in the preparation of hexavalent Americium (Am) during the separation of Am from the lanthanides., hydrothermal synthesis of bismuth oxychloride (BiOCl) micrometer-sized single crystals. It is also used as a precursor in the synthesis of bismuth nanoforms under microwave (MW) irradiation. It used to converts almost any manganese compound to permanganate, which is easily assayed spectrophotometrically.

Solubility
Soluble in aqueous acid. Insoluble in water.

Notes
Hygroscopic. Store in cool, dry place in tightly closed containers. Store away from oxidizing agent and water/moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12232-99-4
Molecular Formula BiNaO3
Molecular Weight (g/mol) 279.967
MDL Number MFCD00003474
InChI Key PNYYBUOBTVHFDN-UHFFFAOYSA-N
Synonym sodium bismuthate, bismuth sodium oxide, sodium bismuthate v, bismuth sodium trioxide, bismuthate bio31-, sodium, sodium oxido dioxo bismuth, sodium bismuth oxide, bio3.na, bismuthate bio31#-, sodium
PubChem CID 4063671
IUPAC Name sodium;oxido(dioxo)bismuth
SMILES [O-][Bi](=O)=O.[Na+]

Specifications

Quantity 25 g
Merck Index 14,8585
Solubility Information Soluble in aqueous acid. Insoluble in water.
Formula Weight 279.97
Percent Purity 80%
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Sodium bismuth oxide
compliance-icons
Product Identifier
  • Sodium bismuth oxide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.