missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sodium Biselenite

Product Code. 10679634
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10679634 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10679634 Supplier Thermo Scientific™ Supplier No. LP0121A
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Prepare Selenite Broth Base, Mannitol Selenite Broth Base, Selenite Cystine Broth Base and Mannitol Selenite Broth Base with Sodium Biselenite.

Use Thermo Scientific™ Oxoid™ Sodium Biselenite in preparation of Selenite Broth Base (CM0395), Mannitol Selenite Broth Base (CM0399) or Selenite Cystine Broth Base (CM0699). The complete medium is recommended for selective enrichment of Salmonella species from various samples.

Dissolve 4 grams in 1 liter of distilled water and use this solution to reconstitute the base medium or tablets.

Toxic by inhalation and if swallowed. Danger of cumulative effects.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 10°C to 30°C
Description Sodium Biselenite
Product Type Sodium Biselenite
Quantity 100 g
compliance-icons compliance-icons compliance-icons
Product Identifier
  • SODIUM BISELENITE (L121)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H331-Toxic if inhaled.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P330-Rinse mouth.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contain: Sodium hydrogen selenite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.