missing translation for 'onlineSavingsMsg'
Learn More

Sodium bis(trifluoromethylsulfonyl)imide

Product Code. 15472727
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15472727

Brand: Thermo Scientific Alfa Aesar 046640.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 91742-21-1 | C2HF6NNaO4S2 | 304.133 g/mol

Sodium bis(trifluoromethylsulfonyl)imide is used as an electrolyte in batteries and fuel cells. It also finds use as an ionic liquid and Lewis acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91742-21-1
Molecular Formula C2HF6NNaO4S2
Molecular Weight (g/mol) 304.133
MDL Number MFCD14156021
InChI Key QXZNUMVOKMLCEX-UHFFFAOYSA-N
Synonym Sodium triflimide; Sodium trifluoromethanesulfonimide
PubChem CID 131887538
IUPAC Name sodium;1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide
SMILES C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F.[Na]

Specifications

Melting Point 257°C to 258°C
Linear Formula (CF3SO2)2NNa
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 303.13
Physical Form Solid
Chemical Name or Material Sodium bis (trifluoromethylsulfonyl)imide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.