missing translation for 'onlineSavingsMsg'
Learn More

Sodium bis(2-methoxyethoxy)aluminum hydride, 70 wt% sol. in toluene (ca. 3.5M)

Product Code. 10686202 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
50g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10686202

Brand: Thermo Scientific Acros 187902500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 22722-98-1 | C6H16AlNaO4

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22722-98-1, 108-88-3
Molecular Formula C6H16AlNaO4

Specifications

Color Amber to Colorless
Boiling Point 110.0°C
Linear Formula [(CH3OCH2CH2O)2AlH2]Na
Fieser 02,382; 03,260; 04,441; 05,596; 06,528; 07,327; 08,448; 12,440; 15,290
Density 1.0360g/mL
Solubility Information Solubility in water: reacts. Other solubilities: soluble in aromatic hydrocarbons and ethers
Formula Weight 202.16
Percent Purity 68.5 to 71.5%
Packaging Glass bottle
Specific Gravity 1.036
Viscosity 65 mPa.s (20°C)
Quantity 250 g
Flash Point 4°C
Physical Form Viscous Liquid
Chemical Name or Material Sodium bis(2-methoxyethoxy)Aluminum hydride
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium bis(2-methoxyethoxy)aluminiumhydride, 70 wt% sol. in toluene (ca. 3.5M)
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.