Learn More
Sodium biphenyl, 15-25%w/w solution in diethylene glycol diethyl ether, offered as 20 x 15mL, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 419423000
98.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : 29
Additional Details : CAS Number : 112-36-7 Weight : 0.30090kg Transport : UN number : 2920 Chem class : 8 Pack group : II
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
Sodium biphenyl, 20%w/w solution in diethylene glycol diethyl ether | |
20%w/w Solution in Diethylene Glycol Diethyl Ether | |
C12H10Na | |
MFCD00013182 | |
sodium biphenyl, diphenyl sodium, acmc-1b11a | |
C1=CC=C(C=C1)C2=CC=CC=C2.[Na] | |
177.202 | |
177.2 | |
Reagent | |
300 mL | |
Solution |
Black-Blue to Green | |
5137-46-2,112-36-7 | |
C6H5C6H5·Na | |
1.0030g/mL | |
AEBYJSOWHQYRPK-UHFFFAOYSA-N | |
1,1'-biphenyl;sodium | |
53446764 | |
17 to 23% (w/w) (typically), actual strength on certificate of analysis | |
1.003 | |
54°C |
Safety and Handling
- Sodium biphenyl, solution in diethylene glycol diethyl ether, 10-23%
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H226-Flammable liquid and vapour.
- H411-Toxic to aquatic life with long lasting effects.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only