missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Bicarbonate USP GenAR Powder, Macron Fine Chemicals™
Click to view available options
:
Poly; 2.5kg
Specifications
Specifications
| CAS | 144-55-8 |
| Physical Form | Powder |
| Quantity | 2.5 kg |
| Molecular Formula | CHNaO3 |
| MDL Number | MFCD00003528 |
| Synonym | sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut |
| InChI Key | UIIMBOGNXHQVGW-UHFFFAOYSA-M |
| SMILES | [Na+].OC([O-])=O |
| IUPAC Name | sodium hydrogen carbonate |
| Molecular Weight (g/mol) | 84.01 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction