missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bicarbonate, Powder, Multi-Compendial, U.S.P., J.T. Baker™

Product Code. 15297246 Shop All J.T.Baker Products
Click to view available options
Quantity:
2.5 kg
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
2.5kg
This item is not returnable. View return policy

Product Code. 15297246

Brand: Avantor J.T.Baker 351005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 144-55-8
Physical Form Powder
Quantity 2.5 kg
Molecular Formula CHNaO3
MDL Number MFCD00003528
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
SMILES [Na+].OC([O-])=O
IUPAC Name sodium;hydrogen carbonate
Molecular Weight (g/mol) 84.01
PubChem CID 516892
ChEBI CHEBI:32139
Formula Weight 84.01
Grade Multi-Compendial/USP
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.20kg
Chemical Name or Material Sodium Bicarbonate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.