missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium bicarbonate, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Unit Size:
12kg
1kg
2.5kg
25g
500g
50kg
5kg
Pack of 4
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 1 kg |
| Linear Formula | NaHCO3 |
| UN Number | NONH for all modes of transport |
| Beilstein | 4153970 |
| IUPAC Name | sodium;hydrogen carbonate |
| Formula Weight | 84.01g/mol |
| Percent Purity | 99.7 to 100.3% |
| Chemical Name or Material | Sodium bicarbonate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction