missing translation for 'onlineSavingsMsg'
Learn More

Sodium Azide, TRC

Product Code. 30006202
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006202 5 g 5g
30003881 25 g 25g
30006259 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006202 Supplier Toronto Research Chemicals Supplier No. TRCS6125305G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26628-22-8
Molecular Formula N3 . Na
Molecular Weight (g/mol) 65.01
Synonym Sodium Azide, Agrizide, Hydrazoic Acid Sodium Salt, Micro-O-Protect, NSC 3072, Nemazyd, SEP 100
IUPAC Name sodium;azide
SMILES [Na+].[N-]=[N+]=[N-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/N3.Na/c1-3-2;/q-1;+1
Formula Weight 64.999
Chemical Name or Material Sodium Azide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.