missing translation for 'onlineSavingsMsg'
Learn More

Sodium Azide, MP Biomedicals

Product Code. 11485985 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485985 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11485985 Supplier MP Biomedicals Supplier No. 0210289110
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Bacteriostatic agent commonly used as a preservative for laboratory reagents and culture media.

  • Sodium azide used as a preservative for laboratory reagents; a bacteriostatic agent in storage solutions
  • Inhibits gram negative flora so is therefore included in culture media for the selective isolation of Streptococci and Staphylococci
  • It has been used to avoid bacterial contamination during RPC-5 column chromatography of DNA fragments

Specifications

Melting Point >300°C (decomposition)
CAS 26628-22-8
Color White
Physical Form Crystalline
Quantity 10 g
Molecular Formula N3Na
Linear Formula NaN3
MDL Number MFCD00003536
Synonym sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
Solubility Information Soluble in water (100mg/mL - clear, colorless solution); slightly soluble in alcohol and ethanol; insoluble in ether.
InChI Key PXIPVTKHYLBLMZ-UHFFFAOYSA-N
SMILES [N-]=[N+]=[N-].[Na+]
IUPAC Name sodium;azide
Molecular Weight (g/mol) 65.011
PubChem CID 33557
ChEBI CHEBI:278547
Formula Weight 65
Percent Purity ≥99%
Chemical Name or Material SODIUM AZIDE
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium Azide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301-IF SWALLOWED:.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.