missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
N3Na, CAS Number-26628-22-8, azoture de sodium, hydrazoic acid, sodium salt, azide, sodium, sodium azide, kazoe, smite, nemazyd, natriumazid german, sodium azide na n3, natriumazid, 100g, 300 deg.C, CHEBI:278547, White, 65.01g/mol, PXIPVTKHYLBLMZ-UHFFFAOYSA-N, sodium;azide, 275 deg.C, 3536, 65.011
Specifications
Specifications
| Boiling Point | 300°C |
| Melting Point | 275°C |
| Color | White |
| pH | 10 |
| Physical Form | Powder |
| Quantity | 100 g |
| Formula Weight | 65.01g/mol |
| Grade | Extra Pure |
| Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Sodium azide
- Danger
- Acute toxicity Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H300-Fatal if swallowed.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P273-Avoid release to the environment.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?