missing translation for 'onlineSavingsMsg'
Learn More

Sodium aurothiomalate(I), 99.9% (metals basis)

Product Code. 11344707
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344707 1 g 1g
11354707 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11344707 Supplier Thermo Scientific Alfa Aesar Supplier No. 039740.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 12244-57-4 | C4H5AuNa2O5S | 408.09 g/mol

Sodium aurothiomalate(I) is useful for high mobility group box chromosomal protein 1 western blotting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12244-57-4
Molecular Formula C4H5AuNa2O5S
Molecular Weight (g/mol) 408.09
MDL Number MFCD00064304,MFCD00064304
InChI Key YLQOAPBVYJCTPW-UHFFFAOYNA-K
Synonym gold sodium thiomalate
PubChem CID 133108869
IUPAC Name gold;sodium;2-sulfanylbutanedioic acid
SMILES O.[Na+].[Na+].[Au+].[O-]C(=O)CC([S-])C([O-])=O

Specifications

Odor Sulphurous
Quantity 1 g
Merck Index 14,4518
Solubility Information Very soluble in water; Practically insoluble in alcohol, ether
Formula Weight 390.08
Percent Purity 99.9%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Sodium aurothiomalate(I)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.