missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium arsenite, 0.1N Standardized Solution, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
4 L
Description
Sodium arsenite is used as hide preservative, antiseptic, dyeing and soaps. It is used as a stressor to induce production of heat shock proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 500 mL |
Assay Percent Range | 0.1N Standardized Solution |
Linear Formula | NaAsO2 |
UN Number | UN1686 |
Solubility Information | Miscible with water. Slightly miscible with alcohol. |
Formula Weight | 129.91 |
Concentration or Composition (by Analyte or Components) | 0.1N |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'