missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium antimonate trihydrate, 98+%, Thermo Scientific™
Click to view available options
Quantity:
250 g
Unit Size:
250g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
A fine powder of sodium antimonate trihydrate, a slightly soluble salt of the highest available was used to prepare a dilute solution of sodium antimonate. In polyol method it is found that the ferrite is impossibly gained when diethylene glycol is replaced by ethylene glycol, or anhydrous sodium acetate by acetic acid sodium salt trihydrate.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from incompatible materials such as strong oxidizing agents, strong acids, strong bases.
A fine powder of sodium antimonate trihydrate, a slightly soluble salt of the highest available was used to prepare a dilute solution of sodium antimonate. In polyol method it is found that the ferrite is impossibly gained when diethylene glycol is replaced by ethylene glycol, or anhydrous sodium acetate by acetic acid sodium salt trihydrate.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from incompatible materials such as strong oxidizing agents, strong acids, strong bases.
Specifications
Specifications
| Quantity | 250 g |
| UN Number | UN1549 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 246.80 (192.75 Anhydrous) |
| Percent Purity | ≥98% |
| Odor | Odorless |
| Chemical Name or Material | Sodium antimonate trihydrate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction