missing translation for 'onlineSavingsMsg'
Learn More

Sodium antimonate trihydrate, 98+%, Thermo Scientific™

Product Code. 11493187
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493187 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11493187 Supplier Thermo Scientific Alfa Aesar Supplier No. A16141.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
A fine powder of sodium antimonate trihydrate, a slightly soluble salt of the highest available was used to prepare a dilute solution of sodium antimonate. In polyol method it is found that the ferrite is impossibly gained when diethylene glycol is replaced by ethylene glycol, or anhydrous sodium acetate by acetic acid sodium salt trihydrate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from incompatible materials such as strong oxidizing agents, strong acids, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15432-85-6
Molecular Formula NaSbO3·3H2O
MDL Number MFCD00058775

Specifications

Quantity 250 g
UN Number UN1549
Solubility Information Slightly soluble in water.
Formula Weight 246.80 (192.75 Anhydrous)
Percent Purity ≥98%
Odor Odorless
Chemical Name or Material Sodium antimonate trihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.