missing translation for 'onlineSavingsMsg'
Learn More

Sodium Ammonium Phosphate Tetrahydrate (Crystalline/Certified), Fisher Chemical

Product Code. 12966294 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12966294 500 g Glass Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12966294 Supplier Fisher Chemical Supplier No. S 218 500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 79°C
CAS 7783-13-3
Color Colorless
pH 8
Physical Form Solid
Quantity 500 g
Molecular Formula H13NNaO8P
Linear Formula NaNH4HPO4·4H2O
MDL Number MFCD00066970
Synonym unii-k78u8wlv4h, ammonium sodium phosphate tetrahydrate, k78u8wlv4h, phosphorsalz, acmc-20ajp4, ammonium sodium phosphate tetrahydrate mi, sodium monoammonium monohydrogen phosphate tetrahydrate, phosphoric acid, monoammonium monosodium salt, tetrahydrate, salt of phosphorus microcosmic salt, sodium ammonium phosphate tetrahydrate
InChI Key IQTQISLCLWJRPM-UHFFFAOYSA-M
SMILES [NH4+].O.O.O.O.OP(=O)([O-])[O-].[Na+]
IUPAC Name azanium;sodium;hydrogen phosphate;tetrahydrate
Molecular Weight (g/mol) 209.067
PubChem CID 56924540
Formula Weight 209.07
Grade Certified
Identification Pass Test
Packaging Glass Bottle
Density 1.54g/cm³
Chemical Name or Material Sodium Ammonium Phosphate Tetrahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.