Learn More
Sodium amide, 99%, extra pure, Thermo Scientific Chemicals
£17.30 - £42.00
Chemical Identifiers
CAS | 7782-92-5 |
---|---|
Molecular Formula | H2NNa |
Molecular Weight (g/mol) | 39 |
MDL Number | MFCD00011117 |
InChI Key | ODZPKZBBUMBTMG-UHFFFAOYSA-N |
Synonym | sodium amide, sodamide, sodium amide na nh2, unii-5db3g6px9d, 5db3g6px9d, sodium azanide, sodium amide, 50 wt. % suspension in toluene, sodiumamide, natriumamid, sodioamine |
PubChem CID | 24533 |
IUPAC Name | sodium;azanide |
SMILES | [NH2-].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10607101
|
Thermo Scientific Acros
339240250 |
25 g | Glass bottle |
£17.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10498862
|
Thermo Scientific Acros
339241000 |
100 g | Glass bottle |
£42.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7782-92-5 | |
39 | |
ODZPKZBBUMBTMG-UHFFFAOYSA-N | |
24533 | |
[NH2-].[Na+] |
H2NNa | |
MFCD00011117 | |
sodium amide, sodamide, sodium amide na nh2, unii-5db3g6px9d, 5db3g6px9d, sodium azanide, sodium amide, 50 wt. % suspension in toluene, sodiumamide, natriumamid, sodioamine | |
sodium;azanide |
Specifications
400.0°C | |
7782-92-5 | |
Powder, Irregular Flakes or Granules | |
H2NNa | |
MFCD00011117 | |
15, 8714 | |
Solubility in water: reacts violently with water. Other solubilities: decomposes in hot alcohol | |
[NH2-].[Na+] | |
39 | |
39 | |
Extra Pure |
210.0°C | |
Gray-White to White | |
99% | |
NaNH2 | |
01,1034; 02,373; 04,439; 05,591; 06,525; 08,446; 15,288 | |
sodium amide, sodamide, sodium amide na nh2, unii-5db3g6px9d, 5db3g6px9d, sodium azanide, sodium amide, 50 wt. % suspension in toluene, sodiumamide, natriumamid, sodioamine | |
ODZPKZBBUMBTMG-UHFFFAOYSA-N | |
sodium;azanide | |
24533 | |
99% | |
Sodium amide, 99% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Very toxic to aquatic life.
Reacts violently with water.
May form explosive peroxides.<
GHS P Statement
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-971-0
missing translation for 'tsca' : TSCA
- Sodium amide
Signal Word
- Danger
Hazard Category
- Substance which\, in contact with water\, emits flammable gases Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H314-Causes severe skin burns and eye damage.
- H400-Very toxic to aquatic life.
- EUH019-May form explosive peroxides.
- EUH014-Reacts violently with water.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P223-Do not allow contact with water.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
RUO – Research Use Only