missing translation for 'onlineSavingsMsg'
Learn More

Sodium Alginate, MP Biomedicals

Product Code. 11412856 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity unitSize
11412856 100 g 100g
1 options
This item is not returnable. View return policy

Product Code. 11412856

Brand: MP Biomedicals 0521829580

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 9005-38-3
Physical Form Powder
Molecular Formula (C6H7O7)A(C6H7O7)BNa
Quantity 100 g
Chemical Name or Material Sodium Alginate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.