missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetylide, 18 wt.% slurry in xylene

Product Code. 10489861 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10489861 100 g Glass bottle 100g
10591734 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10489861 Supplier Thermo Scientific Acros Supplier No. 342031000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1066-26-8 | C2HNa | 48.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1066-26-8 , 1330-20-7
Molecular Formula C2HNa
Molecular Weight (g/mol) 48.02
MDL Number MFCD00008568
InChI Key SFDZETWZUCDYMD-UHFFFAOYSA-N
Synonym sodium acetylide, sodium ethynide, ethynyl sodium, sodium acetylide suspension, sodium acetylide na c2h, sodium ethyne, monosodium acetylide, slbnqjagykqcki-uhfffaoysa-n, sodium acetylide suspension, 18 wt. % slurry in xylene: light mineral oil purity of contained acetylide
PubChem CID 2733336
ChEBI CHEBI:55387
IUPAC Name sodium;ethyne
SMILES C#[C-].[Na+]

Specifications

Color Brown to Yellow
Density 0.9360g/mL
Flash Point 25°C
Packaging Glass bottle
Linear Formula HC≡CNa
Quantity 100 g
Specific Gravity 0.936
Solubility Information Solubility in water: reacts.
Formula Weight 48.02
Percent Purity 17.0 to 22.0 wt%
Physical Form Slurry
Chemical Name or Material Sodium acetylide, 18wt% slurry in xylene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium acetylide, 18 wt% slurry in xylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H226-Flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.