missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate Trihydrate, MP Biomedicals

Product Code. 11490166 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490166 500 g 500g
11430286 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490166 Supplier MP Biomedicals Supplier No. 02300184.5
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

  • To 800-900 mL deionized water add: 8.20 g sodium acetate and mix
  • Adjust to pH 5.2 with 1.0 M NaOH
  • QS to 1 L with deionized water
  • Mix

Chemical Identifiers

CAS 6131-90-4
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.079
MDL Number MFCD00071557
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
PubChem CID 23665404
ChEBI CHEBI:32138
IUPAC Name sodium;acetate;trihydrate
SMILES CC(=O)[O-].O.O.O.[Na+]

Specifications

Melting Point 56°C to 64°C (literature)
Color White
Boiling Point 123°C (-3H)
Linear Formula CH3COONa . 3H2O
Quantity 500 g
Solubility Information Soluble in water (20 g plus 150 mL).(Lit.)
Formula Weight 136.08
Grade HPLC
Physical Form Crystalline Powder
Chemical Name or Material SODIUM ACETATE TRIHYDRATE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.