missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate Trihydrate, Certified AR for Analysis, Crystal, Fisher Chemical™

Product Code. 10615122 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
2.5 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
2.5kg
500g
5kg
This item is not returnable. View return policy

Product Code. 10615122

Brand: Fisher Chemical S/2040/62

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6131-90-4 | C2H9NaO5 | 136.079 g/mol

Sodium Acetate Trihydrate, Certified AR for Analysis, Crystal

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6131-90-4
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.079
MDL Number 71557
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
PubChem CID 23665404
ChEBI CHEBI:32138
IUPAC Name sodium;acetate;trihydrate
SMILES CC(=O)[O-].O.O.O.[Na+]

Specifications

Melting Point 58°C
Color White
pH 8 to 9.5
Packaging Plastic powder jar
Quantity 2.5 kg
Formula Weight 136.08g/mol
Physical Form Solid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.