missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate Trihydrate, Certified AR for Analysis, Crystal, Fisher Chemical™

£65.20 - £440.00

Chemical Identifiers

CAS 6131-90-4
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.079
MDL Number 71557
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
PubChem CID 23665404
ChEBI CHEBI:32138
IUPAC Name sodium;acetate;trihydrate
SMILES CC(=O)[O-].O.O.O.[Na+]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10030350
View Documents Promotion Details
Fisher Chemical
S/2040/53
500 g Plastic powder jar
£65.20
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10615122
View Documents Promotion Details
Fisher Chemical
S/2040/62
2.5 kg Plastic powder jar
£245.00
2.50kg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10376000
View Documents Promotion Details
Fisher Chemical
S/2040/65
5 kg Plastic keg
£440.00
5kg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sodium Acetate Trihydrate, Certified AR for Analysis, Crystal

Specifications

Chemical Identifiers

6131-90-4
136.079
AYRVGWHSXIMRAB-UHFFFAOYSA-M
23665404
sodium;acetate;trihydrate
C2H9NaO5
71557
sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
CHEBI:32138
CC(=O)[O-].O.O.O.[Na+]

Specifications

6131-90-4
White
C2H9NaO5
sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
CC(=O)[O-].O.O.O.[Na+]
136.079
CHEBI:32138
Solid
58°C
8 to 9.5
71557
AYRVGWHSXIMRAB-UHFFFAOYSA-M
sodium;acetate;trihydrate
23665404
136.08g/mol
Videos
SDS
Documents

Documents

Product Certifications