missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Acetate solution EU Pharmacopoeia 0.2N, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1000 mL
Packaging:
Botle
Unit Size:
1000mL
Specifications
Specifications
| CAS | 6131-90-4 |
| Packaging | Botle |
| Molecular Formula | C2H9NaO5 |
| Quantity | 1000 mL |
| Synonym | sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn |
| InChI Key | AYRVGWHSXIMRAB-UHFFFAOYSA-M |
| SMILES | CC(=O)[O-].O.O.O.[Na+] |
| IUPAC Name | sodium;acetate;trihydrate |
| Molecular Weight (g/mol) | 136.079 |
| PubChem CID | 23665404 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction