missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate hydrate, Puratronic™, 99.9985% (metals basis)

Product Code. 11368828
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368828 100 g 100g
11358828 5 g 5g
11348828 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11368828 Supplier Thermo Scientific Alfa Aesar Supplier No. 041685.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 41484-91-7 | C2H3NaO2 | 82.03 g/mol

Sodium acetate hydrate is used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41484-91-7
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00071557
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate hydrate, sodium acetate monohydrate, acetic acid, sodium salt, monohydrate, c2h3o2.na.h2o, naoac water, water sodium acetate, naoac h2o, naoac.h2o, acmc-20ajp8, sodium hydroxide acetic acid
PubChem CID 23681166
SMILES [Na+].CC([O-])=O

Specifications

Melting Point 58°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.9985%
Solubility Information Soluble in water.(726g/L)at 0 C.
IUPAC Name sodium;acetate;hydrate
Formula Weight 82.03 (Anhydrous)
Odor Odorless
Appearance White crystals
Density 1.45
Chemical Name or Material Sodium acetate hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.